Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080453.2 | NP_001073922.2 | 43 | 6091 | ||
XM_011515260.1 | XP_011513562.1 | 43 | 6091 | |||
XM_017011959.1 | XP_016867448.1 | 43 | 6091 | |||
XM_017011960.1 | XP_016867449.1 | 43 | 6091 | |||
GenBank mRNA | AB037861.1 | - | 25 | 3551 | ||
AL050110.1 | - | 10 | 1341 | |||
AL137358.1 | - | 3 | 243 | |||
AY358482.1 | - | 18 | 2552 | |||
BC004286.1 | - | 24 | 3264 | |||
BC013367.2 | - | 13 | 1847 | |||
BC018777.1 | - | 11 | 1514 | |||
BC069262.1 | - | 15 | 2121 |