Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129887.1 | NP_001123359.1 | 11 | 1360 | ||
NM_014377.1 | NP_055192.1 | 13 | 1519 | |||
XM_005250269.2 | XP_005250326.1 | 14 | 1468 | |||
XM_011516028.2 | XP_011514330.1 | 13 | 1919 | |||
XM_011516029.2 | XP_011514331.1 | 14 | 1380 | |||
XM_011516030.2 | XP_011514332.1 | 9 | 720 | |||
XM_017011968.1 | XP_016867457.1 | 14 | 1555 | |||
GenBank mRNA | AK126382.1 | - | 1 | 364 | ||
AL832847.1 | - | 12 | 1303 | |||
BC000859.1 | - | 5 | 363 | |||
BC125057.1 | - | 12 | 1211 | |||
BC139751.1 | - | 13 | 1267 | |||
X98260.1 | - | 11 | 1153 |