Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF503-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024421.1 | 2 | 864 | |||
NR_024422.1 | 2 | 432 | ||||
NR_110299.1 | 2 | 428 | ||||
NR_110300.1 | 2 | 200 | ||||
NR_110301.1 | 2 | 317 | ||||
NR_110302.1 | 2 | 250 | ||||
GenBank mRNA | BC017426.1 | - | 2 | 432 | ||
BC029963.1 | - | 2 | 200 | |||
BE392199.1 | - | 2 | 321 | |||
BI522594.1 | - | 2 | 867 | |||
BJ990170.1 | - | 2 | 432 | |||
DA634630.1 | - | 2 | 252 |