Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042371.2 | NP_001035830.1 | 1 | 252 | ||
GenBank mRNA | AW161386.1 | - | 1 | 252 | ||
BC035985.1 | - | 1 | 216 |