Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135283.1 | 3 | 230 | |||
NR_135284.1 | 2 | 141 | ||||
GenBank mRNA | BC038218.1 | - | 2 | 540 | ||
BG214947.1 | - | 3 | 231 | |||
BX089989.1 | - | 2 | 143 |