Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CISD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136498.1 | NP_001129970.1 | 4 | 1676 | ||
GenBank mRNA | AK097047.1 | - | 3 | 1927 |