Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136503.1 | NP_001129975.1 | 4 | 374 | ||
GenBank mRNA | AF161441.1 | - | 3 | 233 | ||
BC146979.1 | - | 4 | 362 | 9056907 | ||
BC146984.1 | - | 4 | 362 | 9056912 | ||
BM769757.1 | - | 4 | 357 |