Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105378922 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC018791.1 | - | 1 | 1003 |
Gene Symbol: CD58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017002869.1 | XP_016858358.1 | 6 | 12750 |
Gene Symbol: LOC100506459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126833.1 | - | 2 | 2064 | ||
AL832882.1 | - | 1 | 4156 |