Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047519.1 | 4, 5 | 632 | |||
NR_047521.1 | 3, 4 | 479 | ||||
NR_047524.1 | 2, 3 | 295 | ||||
NR_047525.1 | 3, 4 | 346 | ||||
GenBank mRNA | AK026873.1 | - | 4, 5 | 524 | ||
AK311732.1 | - | 3, 4 | 382 | |||
BC042880.1 | - | 3, 4 | 462 | |||
BC102012.1 | - | 4, 5 | 507 | |||
BC102023.1 | - | 4, 5 | 507 | |||
BC107481.1 | - | 2, 3 | 170 | |||
BM459718.1 | - | 4, 5 | 505 | |||
CN361275.1 | - | 3, 4 | 485 | |||
DA223705.1 | - | 3, 4 | 346 | |||
DA721387.1 | - | 3, 4 | 435 | |||
DB282323.1 | - | 3, 4 | 382 | |||
DR005973.1 | - | 4, 5 | 544 |