Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047519.1 | 4 | 473 | |||
NR_047521.1 | 3 | 320 | ||||
NR_047523.1 | 3 | 320 | ||||
NR_047525.1 | 3 | 187 | ||||
GenBank mRNA | AK026873.1 | - | 4 | 365 | ||
AK311121.1 | - | 3 | 244 | |||
AK311732.1 | - | 3 | 223 | |||
BC042880.1 | - | 3 | 303 | |||
BC102012.1 | - | 4 | 348 | |||
BC102023.1 | - | 4 | 348 | |||
BM459718.1 | - | 4 | 346 | |||
CN361275.1 | - | 3 | 326 | |||
DA223705.1 | - | 3 | 187 | |||
DA721387.1 | - | 3 | 276 | |||
DA780029.1 | - | 3 | 274 | |||
DB282323.1 | - | 3 | 223 | |||
DR005509.1 | - | 4 | 362 | |||
DR005973.1 | - | 4 | 385 |