Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SVOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018711.4 | NP_061181.1 | 2 | 474 | ||
GenBank mRNA | BC033587.1 | - | 2 | 274 | 5243017 | |
BC094722.1 | - | 2 | 121 | 4817829 | ||
DA183247.1 | - | 2 | 481 | |||
DA793417.1 | - | 2 | 313 |