Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145531.2 | NP_001139003.1 | 2 | 750 | ||
NM_001302625.1 | NP_001289554.1 | 1 | 94 | |||
XM_017012010.1 | XP_016867499.1 | 2 | 3240 | |||
XM_017012011.1 | XP_016867500.1 | 3 | 3126 | |||
GenBank mRNA | AK093321.1 | - | 2 | 750 | ||
BC112220.1 | - | 1 | 93 | |||
BC112222.1 | - | 1 | 94 | |||
BC143810.1 | - | 1 | 94 |