Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018725.3 | NP_061195.2 | 4 | 293 | ||
XM_005265310.4 | XP_005265367.1 | 4 | 293 | |||
XM_005265311.4 | XP_005265368.1 | 4 | 318 | |||
XM_005265312.4 | XP_005265369.1 | 4 | 349 | |||
XM_011533941.2 | XP_011532243.1 | 4 | 293 | |||
XM_017006807.1 | XP_016862296.1 | 4 | 293 | |||
GenBank mRNA | AF208110.1 | - | 4 | 259 | ||
AF208111.1 | - | 4 | 259 | |||
AF212365.1 | - | 4 | 263 | |||
AF250309.1 | - | 4 | 254 | |||
AK307749.1 | - | 4 | 275 | |||
AY359122.1 | - | 4 | 260 | |||
BC000980.1 | - | 4 | 263 | 2985728 | ||
DA093617.1 | - | 4 | 293 |