Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024075.1 | 12 | 1498 | |||
GenBank mRNA | AF489700.1 | - | 12 | 1498 | ||
AY181245.1 | - | 8 | 1004 |