Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAXX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141969.1 | NP_001135441.1 | 5, 6 | 1680 | ||
NM_001141970.1 | NP_001135442.1 | 5, 6 | 1611 | |||
NM_001254717.1 | NP_001241646.1 | 4, 5 | 1421 | |||
NM_001350.4 | NP_001341.1 | 5, 6 | 1677 | |||
XM_005248860.3 | XP_005248917.1 | 3, 4 | 527 | |||
GenBank mRNA | AB209493.1 | - | 1, 2 | 2145 | ||
AF015956.2 | - | 4, 5 | 1527 | |||
AF039136.1 | - | 5, 6 | 1620 | |||
AF050179.1 | - | 5, 6 | 1588 | |||
AF097742.1 | - | 4, 5 | 1480 | |||
AK223404.1 | - | 5, 6 | 1678 | |||
AK292187.1 | - | 5, 6 | 1678 | |||
AK303767.1 | - | 5, 6 | 1554 | |||
AK303854.1 | - | 4, 5 | 1378 | |||
AK313034.1 | - | 5, 6 | 1620 | |||
BC000220.1 | - | 475 | ||||
BC109073.1 | - | 5, 6 | 1563 | 40034214 | ||
BC109074.1 | - | 5, 6 | 1563 | 40034219 | ||
CR457085.1 | - | 4, 5 | 1473 | |||
HQ436528.1 | - | 4, 5 | 1473 | |||
HQ436529.1 | - | 4, 5 | 1473 |