Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258338.1 | NP_001245267.1 | 1 | 105 | ||
NM_018838.4 | NP_061326.1 | 1 | 105 | |||
GenBank mRNA | AF112208.1 | - | 1 | 125 | ||
AF217092.1 | - | 1 | 84 | |||
AK222619.1 | - | 1 | 89 | |||
BC005936.1 | - | 1 | 94 | 4069306 | ||
BG943528.1 | - | 1 | 90 | |||
BM548072.1 | - | 172 | ||||
BT007220.1 | - | 1 | 72 | |||
BU532909.1 | - | 1 | 92 | |||
DW441120.1 | - | 1 | 119 | |||
U34343.1 | - | 212 |