Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029401.1 | 8 | 1186 | |||
GenBank mRNA | AL137733.1 | - | 8 | 1186 |
Gene Symbol: FAM157B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011518113.2 | XP_011516415.1 | 12 | 2046 |
Gene Symbol: GTF2IP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132119.1 | 5 | 729 | |||
GenBank mRNA | DB081302.1 | - | 5 | 405 | ||
DB156368.1 | - | 5 | 419 |
Gene Symbol: LOC100134822 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL137655.1 | - | 7 | 805 |
Gene Symbol: LOC100288069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033908.1 | 7 | 1077 |