Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012241.1 | NP_001012241.1 | 9 | 1289 | ||
XM_005257298.4 | XP_005257355.1 | 8 | 2941 | |||
XM_005257299.4 | XP_005257356.3 | 7 | 2893 | |||
GenBank mRNA | AK124185.1 | - | 3 | 781 | ||
AK125654.1 | - | 5 | 899 | |||
AK297619.1 | - | 6 | 610 | |||
BC118997.1 | - | 9 | 1212 | 40084759 | ||
BC122543.1 | - | 9 | 1212 | 40084757 | ||
BX647188.1 | - | 3 | 200 | |||
CR749360.1 | - | 9 | 1240 |