Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC50A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122837.1 | NP_001116309.1 | 5 | 564 | ||
NM_001122839.1 | NP_001116311.1 | 5 | 526 | |||
NM_001287586.1 | NP_001274515.1 | 5 | 591 | |||
NM_001287587.1 | NP_001274516.1 | 6 | 708 | |||
NM_001287588.1 | NP_001274517.1 | 5 | 582 | |||
NM_001287589.1 | NP_001274518.1 | 4 | 402 | |||
NM_001287590.1 | NP_001274519.1 | 5 | 721 | |||
NM_001287591.1 | NP_001274520.1 | 6 | 745 | |||
NM_001287592.1 | NP_001274521.1 | 4 | 477 | |||
NM_018845.3 | NP_061333.2 | 6 | 688 | |||
XM_005245339.4 | XP_005245396.1 | 6 | 874 | |||
XM_005245340.3 | XP_005245397.1 | 6 | 955 | |||
XM_005245341.4 | XP_005245398.1 | 6 | 906 | |||
XM_005245342.3 | XP_005245399.1 | 5 | 514 | |||
XM_006711453.2 | XP_006711516.1 | 6 | 706 | |||
GenBank mRNA | AF126023.1 | - | 6 | 648 | ||
AF126024.1 | - | 5 | 513 | |||
AK289551.1 | - | 6 | 689 | |||
BC005943.1 | - | 6 | 632 | 4076256 | ||
BC009621.1 | - | 5 | 488 | 3896154 | ||
BC010869.1 | - | 5 | 479 | |||
BC012378.1 | - | 5 | 513 | |||
BI195885.1 | - | 5 | 723 | |||
BM806886.1 | - | 6 | 732 | |||
BU160680.1 | - | 6 | 630 | |||
BU564973.1 | - | 5 | 569 | |||
BX093804.1 | - | 4 | 394 | |||
CR457148.1 | - | 6 | 588 | |||
X84712.1 | - | 4 | 273 |