Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031710.2 | NP_001026880.2 | 2 | 454 | ||
NM_001172428.1 | NP_001165899.1 | 2 | 454 | |||
NM_018846.4 | NP_061334.4 | 2 | 538 | |||
NR_033328.1 | 3 | 585 | ||||
NR_033329.1 | 3 | 585 | ||||
XM_006715753.2 | XP_006715816.1 | 2 | 449 | |||
XM_006715754.2 | XP_006715817.1 | 3 | 532 | |||
XM_006715755.2 | XP_006715818.1 | 3 | 751 | |||
XM_006715756.2 | XP_006715819.1 | 2 | 620 | |||
XM_006715757.3 | XP_006715820.1 | 2 | 449 | |||
XM_017012439.1 | XP_016867928.1 | 3 | 751 | |||
XM_017012440.1 | XP_016867929.1 | 2 | 449 | |||
XM_017012441.1 | XP_016867930.1 | 3 | 532 | |||
GenBank mRNA | AF111113.1 | - | 2 | 338 | ||
AK056390.1 | - | 3 | 536 | |||
AK290479.1 | - | 2 | 248 | |||
AK297369.1 | - | 379 | ||||
AK297421.1 | - | 2 | 455 | |||
AK299006.1 | - | 2 | 180 | |||
AK302995.1 | - | 3 | 308 | |||
AL136597.1 | - | 2 | 400 | |||
AY028802.1 | - | 2 | 103 | |||
BC009555.1 | - | 3 | 501 | |||
BC039585.1 | - | 2 | 396 | 5527346 | ||
BG704835.1 | - | 2 | 543 | |||
DC370850.1 | - | 2 | 454 | |||
EF560731.1 | - | 3 | 304 |