Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929926 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322244.1 | NP_001309173.1 | 6 | 525 | ||
NR_136239.1 | 6 | 496 | ||||
XM_011512272.1 | XP_011510574.1 | 7 | 609 | |||
XM_011512275.1 | XP_011510577.1 | 7 | 605 | |||
XM_011512276.1 | XP_011510578.1 | 7 | 605 | |||
XM_011512278.1 | XP_011510580.1 | 6 | 521 | |||
XM_011512279.1 | XP_011510581.1 | 5 | 503 | |||
XM_017003148.1 | XP_016858637.1 | 7 | 609 | |||
XM_017003149.1 | XP_016858638.1 | 5 | 507 | |||
XM_017003150.1 | XP_016858639.1 | 6 | 533 | |||
GenBank mRNA | BI461950.1 | - | 4 | 561 | ||
BI828150.1 | - | 6 | 494 | |||
BX113116.1 | - | 6 | 474 |