Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA2012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277372.2 | NP_001264301.2 | 14 | 2358 | ||
XM_006712172.2 | XP_006712235.1 | 14 | 2363 | |||
XM_011510467.2 | XP_011508769.1 | 14 | 2471 | |||
XM_011510468.2 | XP_011508770.1 | 14 | 2366 | |||
XM_017003111.1 | XP_016858600.1 | 14 | 2471 | |||
GenBank mRNA | AB095932.1 | - | 2 | 2866 | ||
AK297413.1 | - | 15 | 2548 | |||
BC118982.1 | - | 2 | 679 | 40026312 |