Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001375.2 | NP_001366.1 | 6 | 1983 | ||
GenBank mRNA | AF047016.1 | - | 6 | 1909 | ||
AF060222.1 | - | 6 | 1930 |