Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018900.3 | NP_061723.1 | 1 | 523 | ||
NM_031410.2 | NP_113598.1 | 1 | 523 | |||
NM_031411.2 | NP_113599.1 | 1 | 523 | |||
GenBank mRNA | AF152305.1 | - | 1 | 368 | ||
AF152479.1 | - | 1 | 368 | |||
AF169695.1 | - | 1 | 368 | |||
DA330326.1 | - | 1 | 523 |