Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080468.2 | NP_001073937.1 | 2 | 421 | ||
GenBank mRNA | AK130951.1 | - | 2 | 432 | ||
BC121075.2 | - | 2 | 425 | 40119665 | ||
BC121076.2 | - | 2 | 425 | 40119666 |