Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038705.1 | NP_001033794.1 | 4 | 2241 | ||
GenBank mRNA | DQ318360.1 | - | 4 | 2241 |
Set Membership: |
G-protein coupled receptor |