Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077706.2 | NP_001071174.1 | 10 | 1433 | ||
NM_001195037.2 | NP_001181966.1 | 9 | 1293 | |||
XM_006715472.3 | XP_006715535.1 | 10 | 1426 | |||
XM_011535795.2 | XP_011534097.1 | 9 | 1297 | |||
XM_011535797.2 | XP_011534099.1 | 7 | 1009 | |||
XM_017010828.1 | XP_016866317.1 | 10 | 1399 | |||
XM_017010829.1 | XP_016866318.1 | 10 | 1426 | |||
XM_017010830.1 | XP_016866319.1 | 10 | 1426 | |||
GenBank mRNA | AK304425.1 | - | 7 | 991 | ||
BC146884.1 | - | 8 | 1140 | 9056812 | ||
DQ904331.1 | - | 8 | 1087 |