Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LNP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085451.1 | NP_001078920.1 | 4 | 1750 | ||
GenBank mRNA | AK056320.1 | - | 4 | 1453 | ||
BC040562.1 | - | 4 | 1752 | |||
BC067886.1 | - | 4 | 925 | |||
BC126362.1 | - | 4 | 1140 | 8992078 | ||
BC130480.1 | - | 4 | 1140 | 40146511 | ||
BC144031.1 | - | 4 | 1140 | 9052551 |