Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080471.1 | NP_001073940.1 | 11 | 1472 | ||
XM_005245141.3 | XP_005245198.1 | 12 | 1665 | |||
XM_011509510.2 | XP_011507812.1 | 12 | 1604 | |||
XM_011509511.2 | XP_011507813.1 | 11 | 1560 | |||
XM_011509512.2 | XP_011507814.1 | 11 | 1484 | |||
XM_017001234.1 | XP_016856723.1 | 11 | 2730 | |||
XM_017001235.1 | XP_016856724.1 | 11 | 2600 | |||
XM_017001236.1 | XP_016856725.1 | 11 | 1567 | |||
XM_017001237.1 | XP_016856726.1 | 12 | 1811 | |||
XM_017001238.1 | XP_016856727.1 | 12 | 1750 | |||
XM_017001239.1 | XP_016856728.1 | 12 | 1561 | |||
XM_017001240.1 | XP_016856729.1 | 9 | 1129 | |||
XM_017001241.1 | XP_016856730.1 | 9 | 1277 | |||
XM_017001242.1 | XP_016856731.1 | 9 | 1181 | |||
GenBank mRNA | AK074121.1 | - | 6 | 1085 |