Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MROH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288814.1 | NP_001275743.1 | 38 | 4530 | ||
NM_032450.2 | NP_115826.2 | 40 | 4670 | |||
XM_011517270.2 | XP_011515572.1 | 41 | 4870 | |||
XM_011517271.2 | XP_011515573.1 | 40 | 4807 | |||
XM_011517272.2 | XP_011515574.1 | 40 | 4843 | |||
XM_011517273.2 | XP_011515575.1 | 40 | 4806 | |||
XM_011517274.2 | XP_011515576.1 | 39 | 4779 | |||
XM_011517275.2 | XP_011515577.1 | 39 | 4617 | |||
XM_011517276.2 | XP_011515578.1 | 41 | 4896 | |||
XM_011517277.1 | XP_011515579.1 | 41 | 4732 | |||
XM_011517278.1 | XP_011515580.1 | 40 | 4612 | |||
XM_011517279.2 | XP_011515581.1 | 40 | 4776 | |||
XM_011517281.1 | XP_011515583.1 | 40 | 4785 | |||
XM_011517282.1 | XP_011515584.1 | 39 | 4710 | |||
XM_011517283.2 | XP_011515585.1 | 41 | 4691 | |||
XM_017013802.1 | XP_016869291.1 | 39 | 4779 | |||
XM_017013803.1 | XP_016869292.1 | 38 | 4589 | |||
XM_017013804.1 | XP_016869293.1 | 40 | 4705 | |||
XM_017013805.1 | XP_016869294.1 | 39 | 4585 | |||
XM_017013806.1 | XP_016869295.1 | 38 | 4347 | |||
GenBank mRNA | AL834145.1 | - | 23 | 2928 | ||
AL834309.1 | - | 39 | 4554 | |||
BC017118.1 | - | 8 | 1087 | |||
CN333563.1 | - | 1 | 24 |