Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039752.3 | NP_001034841.3 | 2 | 624 | ||
NR_134874.1 | 3 | 379 | ||||
XM_011545016.2 | XP_011543318.1 | 4 | 1629 | |||
XM_017017699.1 | XP_016873188.1 | 3 | 426 | |||
XM_017017700.1 | XP_016873189.1 | 4 | 1186 | |||
XM_017017701.1 | XP_016873190.1 | 4 | 1629 | |||
XM_017017702.1 | XP_016873191.1 | 4 | 1629 | |||
XM_017017704.1 | XP_016873193.1 | 4 | 1185 | |||
GenBank mRNA | AB075876.1 | - | 3 | 379 | ||
AK298430.1 | - | 3 | 426 | |||
BC171744.1 | - | 2 | 283 | |||
BX091233.1 | - | 2 | 624 | |||
DR003989.1 | - | 2 | 158 |