Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720543.3 | XP_006720606.1 | 17 | 4907 | ||
XM_006720544.3 | XP_006720607.1 | 16 | 4883 | |||
XM_017022240.1 | XP_016877729.1 | 18 | 4778 | |||
XM_017022241.1 | XP_016877730.1 | 17 | 4263 | |||
XM_017022243.1 | XP_016877732.1 | 17 | 4259 | |||
XM_017022244.1 | XP_016877733.1 | 17 | 4161 | |||
XM_017022245.1 | XP_016877734.1 | 18 | 4216 | |||
XM_017022246.1 | XP_016877735.1 | 17 | 4317 | |||
XM_017022251.1 | XP_016877740.1 | 10 | 3278 | |||
XM_017022252.1 | XP_016877741.1 | 9 | 3254 | |||
XM_017022253.1 | XP_016877742.1 | 18 | 4949 | |||
XM_017022254.1 | XP_016877743.1 | 18 | 4982 |