Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVPLL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145127.1 | NP_001138599.1 | 11 | 1862 | ||
XM_011523982.2 | XP_011522284.1 | 11 | 1945 | |||
XM_011523985.2 | XP_011522287.1 | 10 | 1858 | |||
XM_011523986.2 | XP_011522288.1 | 3 | 757 | |||
GenBank mRNA | AK298286.1 | - | 11 | 1846 |