Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018972.2 | NP_061845.2 | 1 | 156 | ||
NR_046346.1 | 1 | 156 | ||||
XM_011517551.2 | XP_011515853.1 | 1 | 1288 | |||
XM_011517552.2 | XP_011515854.1 | 1 | 144 | |||
XM_017013585.1 | XP_016869074.1 | 1 | 1288 | |||
XM_017013586.1 | XP_016869075.1 | 1 | 1288 | |||
GenBank mRNA | AB551556.1 | - | 1 | 155 | ||
AB551557.1 | - | 1 | 155 | |||
AB551558.1 | - | 1 | 153 | |||
AK292572.1 | - | 1 | 95 | |||
AK295594.1 | - | 1 | 95 | |||
DB505741.1 | - | 1 | 156 | |||
Y17849.1 | - | 1 | 84 |