Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011530435.2 | XP_011528737.1 | 2 | 251 | ||
XM_011530436.2 | XP_011528738.1 | 3 | 306 | |||
XM_011530437.1 | XP_011528739.1 | 2 | 976 | |||
XM_011530438.2 | XP_011528740.1 | 2 | 492 | |||
XM_017028990.1 | XP_016884479.1 | 2 | 747 |