Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAIAP2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018842.4 | NP_061330.2 | 6 | 705 | ||
GenBank mRNA | AF119666.2 | - | 6 | 658 | ||
AK027109.1 | - | 2 | 160 | |||
AK309936.1 | - | 6 | 680 | |||
BC013888.2 | - | 6 | 658 | 3842949 |