Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161009.2 | NP_001154481.1 | 1, 2 | 531 | ||
NM_001161011.2 | NP_001154483.1 | 1, 2 | 531 | |||
NM_001161012.2 | NP_001154484.1 | 1, 2 | 531 | |||
NM_001161013.2 | NP_001154485.1 | 1, 2 | 531 | |||
GenBank mRNA | AF275808.1 | - | 1, 2 | 39 | ||
AK225082.1 | - | 1, 2 | 264 | |||
AK291818.1 | - | 1, 2 | 259 | |||
AK301565.1 | - | 1, 2 | 264 | |||
AY150851.1 | - | 1, 2 | 531 | |||
AY150853.1 | - | 1, 2 | 531 | |||
BC003586.1 | - | 1, 2 | 251 | 3357276 |