Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018915.3 | NP_061738.1 | 1 | 665 | ||
NM_032009.2 | NP_114398.1 | 1 | 665 | |||
GenBank mRNA | AF152322.1 | - | 1 | 453 | ||
AF152508.1 | - | 1 | 453 | |||
AL536793.3 | - | 1 | 680 | |||
BC093871.1 | - | 1 | 504 | 7939716 | ||
BC104950.1 | - | 1 | 504 | 8143953 |