Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHGA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018921.2 | NP_061744.1 | 1 | 410 | ||
NM_032089.1 | NP_114478.1 | 1 | 410 | |||
GenBank mRNA | AF152329.1 | - | 1 | 410 | ||
AF152516.1 | - | 1 | 410 | |||
BC132769.1 | - | 1 | 431 | 40146791 |