Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278610.1 | NP_001265539.1 | 12 | 2068 | ||
NM_002466.3 | NP_002457.1 | 13 | 2140 | |||
GenBank mRNA | AK223482.1 | - | 13 | 1875 | ||
AK303249.1 | - | 12 | 1973 | |||
AK314791.1 | - | 13 | 2045 | |||
BC007585.1 | - | 13 | 2065 | 3162656 | ||
BC053555.1 | - | 13 | 2034 | 6140320 | ||
BX647151.1 | - | 3 | 2525 | |||
X13293.1 | - | 13 | 2002 |