Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018945.3 | NP_061818.1 | 4 | 559 | ||
XM_005266931.3 | XP_005266988.1 | 3 | 3408 | |||
XM_011535755.2 | XP_011534057.1 | 5 | 388 | |||
XM_017010741.1 | XP_016866230.1 | 3 | 1194 | |||
GenBank mRNA | AB038040.1 | - | 4 | 561 | ||
AB209990.1 | - | 3 | 665 | |||
AJ251860.1 | - | 4 | 553 | |||
AK290582.1 | - | 4 | 592 | |||
BC075082.2 | - | 4 | 282 | 30915597 | ||
BC075083.2 | - | 4 | 286 | 30915385 | ||
EF108327.1 | - | 3 | 667 |