Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007246.2 | NP_001007247.1 | 5 | 622 | ||
NM_018963.4 | NP_061836.2 | 5 | 622 | |||
NM_033656.3 | NP_387505.1 | 5 | 622 | |||
XM_011529611.1 | XP_011527913.1 | 5 | 655 | |||
XM_017028373.1 | XP_016883862.1 | 6 | 427 | |||
XM_017028375.1 | XP_016883864.1 | 5 | 655 | |||
GenBank mRNA | AB080586.1 | - | 5 | 426 | ||
AB080587.1 | - | 5 | 426 | |||
AJ238214.2 | - | 5 | 426 | |||
AJ292465.1 | - | 5 | 406 | |||
AJ292466.1 | - | 5 | 406 | |||
BC064602.1 | - | 5 | 458 | |||
BC142986.1 | - | 5 | 471 |