Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3TC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318480.1 | NP_001305409.1 | 10 | 1421 | ||
NM_018986.4 | NP_061859.3 | 10 | 1338 | |||
NR_134639.1 | 8 | 1052 | ||||
XM_006713889.2 | XP_006713952.1 | 7 | 756 | |||
XM_006713891.3 | XP_006713954.1 | 4 | 398 | |||
XM_011513488.1 | XP_011511790.1 | 10 | 1315 | |||
XM_011513489.2 | XP_011511791.1 | 8 | 938 | |||
XM_017008300.1 | XP_016863789.1 | 10 | 1447 | |||
XM_017008301.1 | XP_016863790.1 | 10 | 1130 | |||
XM_017008302.1 | XP_016863791.1 | 7 | 840 | |||
GenBank mRNA | AK000363.1 | - | 6 | 726 | ||
AK074093.1 | - | 9 | 1177 | |||
AK074402.1 | - | 10 | 1215 | |||
AK126165.1 | - | 10 | 1294 | |||
AK222865.1 | - | 6 | 726 |