Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEM1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018708.2 | NP_061178.1 | 1 | 3323 | ||
GenBank mRNA | AJ311359.1 | - | 1 | 3184 | ||
AK074232.1 | - | 1 | 3287 | |||
AK126532.1 | - | 2 | 3057 | |||
AL359589.1 | - | 1 | 1471 | |||
AL365408.1 | - | 1 | 1785 | |||
AY356352.1 | - | 1 | 3285 | |||
BC004988.1 | - | 1 | 3288 | 2957950 | ||
CR749732.1 | - | 1 | 1471 |