Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR100HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024430.2 | 4 | 930 | |||
NR_137175.1 | 2 | 1201 | ||||
NR_137176.1 | 2 | 912 | ||||
NR_137177.1 | 2 | 891 | ||||
NR_137179.1 | 5 | 884 | ||||
NR_137180.1 | 5 | 942 | ||||
NR_137181.1 | 3 | 827 | ||||
NR_137182.1 | 4 | 909 | ||||
NR_137183.1 | 3 | 818 | ||||
NR_137184.1 | 2 | 736 | ||||
NR_137185.1 | 3 | 797 | ||||
NR_137186.1 | 3 | 706 | ||||
NR_137187.1 | 3 | 685 | ||||
NR_137188.1 | 3 | 775 | ||||
NR_137189.1 | 2 | 1222 | ||||
NR_137190.1 | 3 | 1255 | ||||
NR_137191.1 | 3 | 1234 | ||||
NR_137192.1 | 4 | 1094 | ||||
NR_137193.1 | 5 | 921 | ||||
NR_137194.1 | 3 | 796 | ||||
NR_137195.1 | 2 | 964 | ||||
NR_137196.1 | 2 | 985 | ||||
GenBank mRNA | AB075498.1 | - | 2 | 1454 | ||
AB485963.1 | - | 4 | 813 | |||
AK021552.1 | - | 1 | 96 | |||
AK091713.1 | - | 3 | 586 | |||
AK123947.1 | - | 2 | 1138 | |||
AK311309.1 | - | 4 | 883 | |||
AL832469.1 | - | 2 | 518 | |||
BC040599.1 | - | 2 | 514 | |||
BX538058.1 | - | 2 | 491 | |||
BX647608.1 | - | 2 | 492 | |||
DB248200.1 | - | 3 | 604 |