Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUNDC3A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110802.1 | 4 | 2972 | |||
GenBank mRNA | BC051705.1 | - | 3 | 2731 |