Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01538 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033983.1 | 3 | 606 | |||
GenBank mRNA | BC042493.1 | - | 3 | 604 | ||
BG722547.1 | - | 3 | 609 |