Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHGB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018926.2 | NP_061749.1 | 1 | 1156 | ||
NM_032100.1 | NP_115271.1 | 1 | 1156 | |||
GenBank mRNA | AF152335.1 | - | 1 | 1156 | ||
AF152522.1 | - | 1 | 1156 |