Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC729218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103825.1 | 3 | 2018 | |||
NR_109983.1 | 10 | 2961 | ||||
GenBank mRNA | AK024248.1 | - | 10 | 2932 |
Gene Symbol: LOC101060022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011544347.2 | XP_011542649.1 | 1 | 613 |