Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIS18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018944.2 | NP_061817.1 | 5 | 722 | ||
GenBank mRNA | AF231921.1 | - | 5 | 694 | ||
AF387845.1 | - | 5 | 692 | |||
AK075281.1 | - | 5 | 694 | |||
AK312073.1 | - | 5 | 813 | |||
AW962893.1 | - | 3 | 176 | |||
BC042917.1 | - | 5 | 698 | 5934336 | ||
BU567844.1 | - | 3 | 289 |